Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005981.3 | NP_005972.1 | 4 | 509 | ||
XM_005269074.3 | XP_005269131.2 | 4 | 1434 | |||
XM_017019793.1 | XP_016875282.1 | 2 | 238 | |||
GenBank mRNA | AK090502.1 | - | 2 | 912 | ||
AK294128.1 | - | 4 | 462 | |||
AK315765.1 | - | 4 | 630 | |||
BC010377.1 | - | 4 | 462 | 4294627 | ||
CR407647.1 | - | 4 | 355 | |||
U01160.1 | - | 4 | 457 |