Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256577.2 | NP_001243506.2 | 6 | 1544 | ||
NM_001256580.2 | NP_001243509.2 | 6 | 1599 | |||
NM_001303624.1 | NP_001290553.1 | 6 | 1502 | |||
NM_001303625.1 | NP_001290554.1 | 7 | 1711 | |||
NM_001303626.1 | NP_001290555.1 | 6 | 1366 | |||
NM_006013.4 | NP_006004.3 | 7 | 1707 | |||
GenBank mRNA | BC003358.1 | - | 6 | 1378 | 2900175 | |
BC018570.1 | - | 1 | 249 |
Gene Symbol: DNASE1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW445150.1 | - | 1 | 343 | ||
BE900752.1 | - | 1 | 78 |