Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU2AF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006235.2 | NP_006226.2 | 5 | 2639 | ||
XM_005271593.1 | XP_005271650.1 | 6 | 2781 | |||
XM_005271594.3 | XP_005271651.1 | 6 | 2738 | |||
XM_006718859.1 | XP_006718922.1 | 6 | 2561 | |||
XM_006718860.3 | XP_006718923.1 | 6 | 4639 | |||
XM_017017932.1 | XP_016873421.1 | 5 | 4340 | |||
GenBank mRNA | BC032549.1 | - | 6 | 2426 | 5554134 | |
Z49194.1 | - | 5 | 2911 |