Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270476.1 | NP_001257405.1 | 10, 11 | 1281 | ||
NM_006245.3 | NP_006236.1 | 10, 11 | 1264 | |||
NM_180976.2 | NP_851307.1 | 10, 11 | 1168 | |||
NM_180977.2 | NP_851308.1 | 8, 9 | 946 | |||
GenBank mRNA | AB000634.1 | - | 10, 11 | 1099 | ||
AB000635.1 | - | 8, 9 | 793 | |||
AB451342.1 | - | 10, 11 | 1085 | |||
AB451357.1 | - | 10, 11 | 1085 | |||
AK122701.1 | - | 10, 11 | 1158 | |||
AK290604.1 | - | 10, 11 | 1077 | |||
AK291797.1 | - | 10, 11 | 1172 | |||
AK299696.1 | - | 10, 11 | 1137 | |||
BC001095.2 | - | 10, 11 | 1148 | 3507280 | ||
BC001175.1 | - | 10, 11 | 1075 | 3355395 | ||
BC010692.1 | - | 10, 11 | 1158 | 3853892 | ||
D78360.1 | - | 10, 11 | 1066 | |||
L76702.1 | - | 10, 11 | 1272 |