Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006255.4 | NP_006246.2 | 1 | 855 | ||
XM_011536956.1 | XP_011535258.1 | 1 | 737 | |||
XM_011536957.1 | XP_011535259.1 | 1 | 737 | |||
XM_017021459.1 | XP_016876948.1 | 1 | 737 | |||
GenBank mRNA | AB451313.1 | - | 1 | 196 | ||
AK290183.1 | - | 1 | 574 | |||
AX775933.1 | - | 1 | 584 | |||
BC037268.1 | - | 1 | 584 | 4822159 | ||
CB994021.1 | - | 528 | ||||
M55284.1 | - | 1 | 362 |