Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006265.2 | NP_006256.1 | 5 | 697 | ||
GenBank mRNA | AK289505.1 | - | 5 | 474 | ||
BC050381.1 | - | 5 | 537 | 6044010 | ||
D38551.1 | - | 5 | 593 |