Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042368.2 | NP_001035827.1 | 18 | 3056 | ||
NM_001271774.1 | NP_001258703.1 | 18 | 2968 | |||
NM_001271775.1 | NP_001258704.1 | 18 | 3061 | |||
NM_001271776.1 | NP_001258705.1 | 18 | 3041 | |||
NM_006266.3 | NP_006257.1 | 18 | 3077 | |||
GenBank mRNA | AB037729.1 | - | 11 | 5166 | ||
AF027169.1 | - | 18 | 2972 | |||
AF295773.1 | - | 18 | 2992 | |||
AK000242.1 | - | 13 | 2034 | |||
AK024575.1 | - | 10 | 1631 | |||
AK056462.1 | - | 17 | 2754 | |||
AK074114.1 | - | 18 | 3973 | |||
AK090450.1 | - | 12 | 5027 | |||
AK127524.1 | - | 18 | 3061 | |||
BC021581.1 | - | 6 | 1153 | |||
BC033198.1 | - | 10 | 1473 | |||
BC059362.1 | - | 18 | 3031 | 30340922 | ||
BM805052.1 | - | 3 | 642 |