Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006267.4 | NP_006258.3 | 29 | 10301 | ||
XM_005264002.2 | XP_005264059.1 | 30 | 10438 | |||
XM_005264003.2 | XP_005264060.1 | 30 | 10342 | |||
XM_005264004.2 | XP_005264061.1 | 29 | 10288 | |||
XM_005264005.4 | XP_005264062.1 | 30 | 10426 | |||
XM_005264007.2 | XP_005264064.1 | 30 | 7438 | |||
XM_011511575.2 | XP_011509877.1 | 30 | 10341 | |||
XM_011511576.2 | XP_011509878.1 | 31 | 10189 | |||
XM_011511578.2 | XP_011509880.1 | 30 | 7480 | |||
GenBank mRNA | AK025462.1 | - | 6 | 1804 |