Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006741.3 | NP_006732.3 | 6 | 645 | ||
XM_005268995.4 | XP_005269052.1 | 5 | 469 | |||
XM_006719471.3 | XP_006719534.1 | 4 | 419 | |||
GenBank mRNA | AK123969.1 | - | 5 | 350 | ||
AY063766.1 | - | 6 | 476 | |||
BC022470.1 | - | 6 | 645 | 4819907 | ||
CR457003.1 | - | 6 | 474 | |||
U48707.1 | - | 6 | 482 |