Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078166.1 | NP_001071634.1 | 3 | 1641 | ||
NM_006924.4 | NP_008855.1 | 4 | 1441 | |||
GenBank mRNA | AB062124.1 | - | 4 | 1322 | ||
AB209558.1 | - | 3 | 1526 | |||
AK298527.1 | - | 3 | 1023 | |||
BC010264.1 | - | 4 | 1267 | 2900101 | ||
M69040.1 | - | 4 | 1356 |