Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281786.1 | NP_001268715.1 | 11 | 2007 | ||
NM_004169.4 | NP_004160.3 | 12 | 2123 | |||
NM_148918.2 | NP_683718.1 | 11 | 2006 | |||
XM_005256767.3 | XP_005256824.1 | 13 | 2194 | |||
XM_011523992.2 | XP_011522294.1 | 10 | 1851 | |||
XM_017024957.1 | XP_016880446.1 | 12 | 1948 | |||
XM_017024958.1 | XP_016880447.1 | 12 | 2077 | |||
XM_017024959.1 | XP_016880448.1 | 9 | 1698 | |||
XM_017024960.1 | XP_016880449.1 | 8 | 1575 | |||
GenBank mRNA | AK223552.1 | - | 12 | 2054 | ||
BC007979.2 | - | 12 | 1969 | 3687873 | ||
BC038598.1 | - | 12 | 1980 | 4523709 | ||
BM792235.1 | - | 1 | 250 | |||
Y14486.1 | - | 7 | 2236 |