Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286233.1 | NP_001273162.1 | 16 | 3206 | ||
NM_001286234.1 | NP_001273163.1 | 11 | 2452 | |||
NM_001286235.1 | NP_001273164.1 | 13 | 2750 | |||
NM_001286236.1 | NP_001273165.1 | 10 | 2291 | |||
NM_001286237.1 | NP_001273166.1 | 10 | 2345 | |||
NM_153449.3 | NP_703150.1 | 12 | 2488 | |||
XM_005253315.3 | XP_005253372.1 | 11 | 2286 | |||
XM_005253317.4 | XP_005253374.1 | 11 | 2805 | |||
XM_011520562.1 | XP_011518864.1 | 14 | 2640 | |||
XM_011520563.2 | XP_011518865.1 | 11 | 2300 | |||
XM_011520564.2 | XP_011518866.1 | 10 | 2109 | |||
XM_011520565.2 | XP_011518867.1 | 9 | 2138 | |||
XM_017018841.1 | XP_016874330.1 | 12 | 2385 | |||
XM_017018842.1 | XP_016874331.1 | 15 | 2956 | |||
XM_017018843.1 | XP_016874332.1 | 15 | 3280 | |||
XM_017018844.1 | XP_016874333.1 | 12 | 2335 | |||
XM_017018845.1 | XP_016874334.1 | 12 | 2877 | |||
XM_017018846.1 | XP_016874335.1 | 12 | 2461 | |||
XM_017018847.1 | XP_016874336.1 | 11 | 2418 | |||
GenBank mRNA | AK122783.1 | - | 13 | 2537 | ||
AK126026.1 | - | 16 | 2987 | |||
AL110298.1 | - | 5 | 1531 | |||
AL832448.1 | - | 9 | 2111 | |||
AY357941.1 | - | 11 | 2431 | |||
BC060766.1 | - | 11 | 2299 | 5297510 |
Gene Symbol: SLC2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006931.2 | NP_008862.1 | 10 | 2505 | ||
GenBank mRNA | AB209607.1 | - | 9 | 2983 | ||
AK074861.1 | - | 2 | 1143 | |||
BC039196.1 | - | 10 | 2380 | 4396508 | ||
M20681.1 | - | 10 | 2485 |