Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291916.1 | NP_001278845.1 | 4 | 846 | ||
NM_006938.3 | NP_008869.1 | 4 | 969 | |||
GenBank mRNA | AJ577268.1 | - | 4 | 901 | ||
BC072427.1 | - | 4 | 856 | 6464946 | ||
J03798.1 | - | 4 | 958 |