Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209277.1 | - | 8 | 1091 | ||
AK308978.1 | - | 2 | 115 | |||
BC117261.1 | - | 8 | 1027 | 40125812 | ||
BC143366.1 | - | 8 | 1027 | 9051871 | ||
L13858.1 | - | 8 | 1001 |