Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3IL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC022239.1 | - | 10 | 2276 | 4390573 | |
BC051820.1 | - | 9 | 2117 | 5188805 |