Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278656.1 | NP_001265585.1 | 4 | 1003 | ||
NM_004175.4 | NP_004166.1 | 4 | 1017 | |||
NR_103819.1 | 3 | 824 | ||||
GenBank mRNA | BC034447.1 | - | 4 | 700 | ||
BP202359.1 | - | 3 | 474 | |||
CN286602.1 | - | 4 | 657 |