Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272011.1 | NP_001258940.1 | 6 | 615 | ||
NM_014226.2 | NP_055041.1 | 7 | 705 | |||
XM_006720225.2 | XP_006720288.1 | 7 | 705 | |||
XM_006720226.2 | XP_006720289.1 | 7 | 705 | |||
XM_011537052.1 | XP_011535354.1 | 7 | 705 | |||
XM_017021549.1 | XP_016877038.1 | 7 | 702 | |||
XM_017021550.1 | XP_016877039.1 | 6 | 601 | |||
XM_017021551.1 | XP_016877040.1 | 8 | 645 | |||
XM_017021552.1 | XP_016877041.1 | 7 | 743 | |||
XM_017021553.1 | XP_016877042.1 | 5 | 540 | |||
XM_017021554.1 | XP_016877043.1 | 5 | 539 | |||
XM_017021555.1 | XP_016877044.1 | 7 | 702 | |||
XM_017021556.1 | XP_016877045.1 | 7 | 705 | |||
XM_017021557.1 | XP_016877046.1 | 6 | 605 | |||
XM_017021558.1 | XP_016877047.1 | 6 | 568 | |||
XM_017021559.1 | XP_016877048.1 | 6 | 694 | |||
GenBank mRNA | AB022694.1 | - | 7 | 699 | ||
AK094747.1 | - | 5 | 512 | |||
AK302349.1 | - | 6 | 600 | |||
AK312778.1 | - | 7 | 696 | |||
BC026069.1 | - | 6 | 1116 | 4838296 | ||
BC043179.1 | - | 5 | 1461 | 5287879 | ||
BC053536.1 | - | 7 | 681 | 5175851 |