Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011658.3 | NP_001011658.1 | 6 | 1951 | ||
NM_001128835.2 | NP_001122307.2 | 6 | 1945 | |||
NM_014563.5 | NP_055378.1 | 5 | 1809 | |||
XM_011545565.1 | XP_011543867.1 | 7 | 2019 | |||
XM_011545566.2 | XP_011543868.1 | 6 | 1875 | |||
GenBank mRNA | AK055671.1 | - | 1 | 1116 | ||
BC016915.1 | - | 6 | 1076 | 4432196 | ||
BC052618.1 | - | 5 | 1782 | 6581382 |