Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014946.3 | NP_055761.2 | 17 | 3337 | ||
NM_199436.1 | NP_955468.1 | 16 | 3241 | |||
XM_005264516.4 | XP_005264573.1 | 17 | 3113 | |||
XM_011533067.2 | XP_011531369.1 | 16 | 3326 | |||
XM_017004777.1 | XP_016860266.1 | 16 | 3298 | |||
XM_017004778.1 | XP_016860267.1 | 15 | 3230 | |||
GenBank mRNA | AB029006.1 | - | 16 | 3241 | ||
AJ246001.1 | - | 17 | 3241 | |||
BC150260.1 | - | 16 | 3241 | 9007202 |