Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195226.1 | NP_001182155.1 | 14 | 1884 | ||
NM_016335.4 | NP_057419.4 | 15 | 2299 | |||
GenBank mRNA | AB209472.1 | - | 11 | 4575 | ||
AF010310.1 | - | 4 | 787 | |||
AK001359.1 | - | 6 | 2860 | |||
AK091063.1 | - | 9 | 2464 | |||
BC036534.1 | - | 13 | 3011 | |||
BC068260.1 | - | 14 | 2098 | |||
BC094736.1 | - | 14 | 1929 | |||
U79754.1 | - | 15 | 2299 |
Gene Symbol: LOC102724788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724935.3 | XP_006724998.1 | 14 | 2128 | ||
XM_006724936.3 | XP_006724999.1 | 13 | 2038 | |||
XM_017030166.1 | XP_016885655.1 | 14 | 2225 | |||
XM_017030167.1 | XP_016885656.1 | 13 | 2769 | |||
XM_017030168.1 | XP_016885657.1 | 14 | 1946 | |||
XM_017030169.1 | XP_016885658.1 | 12 | 2504 | |||
XM_017030170.1 | XP_016885659.1 | 13 | 1855 |