Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040142.1 | NP_001035232.1 | 14 | 2548 | ||
NM_001040143.1 | NP_001035233.1 | 13 | 2309 | |||
NM_021007.2 | NP_066287.2 | 14 | 2614 | |||
XM_005246753.3 | XP_005246810.1 | 14 | 2564 | |||
XM_005246755.4 | XP_005246812.1 | 9 | 1649 | |||
XM_017004655.1 | XP_016860144.1 | 14 | 2463 | |||
XM_017004656.1 | XP_016860145.1 | 14 | 2519 | |||
XM_017004657.1 | XP_016860146.1 | 14 | 2416 | |||
XM_017004658.1 | XP_016860147.1 | 14 | 2728 | |||
XM_017004659.1 | XP_016860148.1 | 2 | 1646 | |||
GenBank mRNA | AB208888.1 | - | 14 | 2401 | ||
M94055.1 | - | 14 | 2363 |