Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021129.3 | NP_066952.1 | 7 | 644 | ||
GenBank mRNA | AB026723.1 | - | 7 | 567 | ||
AF092439.1 | - | 7 | 614 | |||
AF108211.1 | - | 7 | 525 | |||
AF119665.1 | - | 7 | 614 | |||
AF154065.1 | - | 7 | 582 | |||
AF217186.1 | - | 7 | 537 | |||
AK058193.1 | - | 7 | 637 | |||
AK127102.1 | - | 1 | 591 | |||
BC001022.2 | - | 7 | 619 | 3345496 | ||
BC061581.1 | - | 7 | 627 | 6526628 | ||
BC105034.1 | - | 7 | 610 | 8144037 | ||
BC105036.1 | - | 7 | 610 | 8144039 | ||
BC107882.1 | - | 7 | 578 | 6495107 | ||
BU520836.1 | - | 7 | 625 | |||
EU794626.1 | - | 7 | 644 |