Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021133.3 | NP_066956.1 | 2 | 800 | ||
GenBank mRNA | AK315231.1 | - | 2 | 751 | ||
BC090934.1 | - | 2 | 784 | 3073811 | ||
BC114433.1 | - | 1 | 666 | 40054016 | ||
BC115697.1 | - | 1 | 666 | 40054014 | ||
CR627369.1 | - | 2 | 798 |