Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139443.1 | NP_001132915.1 | 9 | 2001 | ||
NM_001300786.1 | NP_001287715.1 | 8 | 1920 | |||
NM_001300787.1 | NP_001287716.1 | 9 | 2001 | |||
NM_004183.3 | NP_004174.1 | 10 | 2189 | |||
NR_134580.1 | 10 | 2392 | ||||
XM_005274210.3 | XP_005274267.1 | 10 | 2299 | |||
XM_005274215.3 | XP_005274272.1 | 8 | 1954 | |||
XM_005274216.3 | XP_005274273.1 | 9 | 2314 | |||
XM_005274219.3 | XP_005274276.1 | 8 | 2066 | |||
XM_005274221.3 | XP_005274278.1 | 7 | 1913 | |||
XM_011545229.2 | XP_011543531.1 | 10 | 1739 | |||
XM_011545230.2 | XP_011543532.1 | 9 | 1542 | |||
XM_011545233.2 | XP_011543535.1 | 4 | 1436 | |||
XM_017018228.1 | XP_016873717.1 | 8 | 1954 | |||
XM_017018229.1 | XP_016873718.1 | 10 | 2502 | |||
XM_017018230.1 | XP_016873719.1 | 8 | 2157 | |||
GenBank mRNA | AF057169.1 | - | 10 | 1713 | ||
AF057170.1 | - | 10 | 1916 | |||
AF073501.1 | - | 10 | 1697 | |||
AK295998.1 | - | 9 | 1535 | |||
AK298747.1 | - | 5 | 1248 | |||
AY515704.1 | - | 9 | 1609 | |||
BC015220.2 | - | 8 | 1920 | 3877806 | ||
BC041664.1 | - | 9 | 1515 | 5194649 | ||
JQ954696.1 | - | 9 | 1675 | |||
KU178390.1 | - | 7 | 1348 | |||
KU178391.1 | - | 8 | 1429 | |||
KU178392.1 | - | 7 | 1276 | |||
KU178393.1 | - | 3 | 810 | |||
KU178394.1 | - | 3 | 654 |