Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP1GDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100426.1 | NP_001093896.1 | 8 | 991 | ||
NM_001100427.1 | NP_001093897.1 | 8 | 988 | |||
NM_001100428.1 | NP_001093898.1 | 7 | 844 | |||
NM_001100429.1 | NP_001093899.1 | 7 | 841 | |||
NM_001100430.1 | NP_001093900.1 | 6 | 715 | |||
NM_021159.4 | NP_066982.3 | 8 | 991 | |||
GenBank mRNA | AF215923.1 | - | 7 | 798 | ||
AF237413.1 | - | 7 | 798 | |||
AK055032.1 | - | 8 | 911 | |||
AK295156.1 | - | 7 | 803 | |||
AK297543.1 | - | 5 | 538 | |||
AK311243.1 | - | 5 | 526 | |||
AY376900.1 | - | 5 | 525 | |||
BC001816.2 | - | 8 | 898 | 3010166 | ||
BC001851.2 | - | 8 | 898 | 3010166 | ||
BC098269.1 | - | 8 | 849 | 40001059 | ||
BC098334.1 | - | 7 | 699 | 40001057 | ||
BC099708.1 | - | 7 | 699 | 40001058 | ||
BC099845.1 | - | 7 | 702 | 40001061 | ||
BT006837.1 | - | 7 | 801 | |||
X63465.1 | - | 6 | 651 |