Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199797.1 | NP_001186726.1 | 4 | 627 | ||
NM_002832.3 | NP_002823.3 | 4 | 1473 | |||
NM_080588.2 | NP_542155.1 | 3 | 966 | |||
NR_037663.1 | 4 | 1420 | ||||
NR_037664.1 | 3 | 1289 | ||||
XM_011509819.1 | XP_011508121.1 | 4 | 808 | |||
XM_011509820.2 | XP_011508122.1 | 4 | 512 | |||
XM_011509821.1 | XP_011508123.1 | 4 | 440 | |||
XM_011509822.2 | XP_011508124.1 | 4 | 311 | |||
XM_011509823.1 | XP_011508125.1 | 4 | 808 | |||
XM_011509824.1 | XP_011508126.1 | 3 | 316 | |||
XM_017001935.1 | XP_016857424.1 | 4 | 805 | |||
XM_017001936.1 | XP_016857425.1 | 4 | 805 | |||
XM_017001937.1 | XP_016857426.1 | 4 | 623 | |||
GenBank mRNA | AF394064.1 | - | 2 | 410 | ||
AK127214.1 | - | 4 | 1466 | |||
AK225920.1 | - | 4 | 471 | |||
AK293186.1 | - | 3 | 316 | |||
AK301051.1 | - | 4 | 441 | |||
AK302869.1 | - | 4 | 621 | |||
BC001746.1 | - | 3 | 966 | 3543111 | ||
BG340453.1 | - | 4 | 509 | |||
BT009848.1 | - | 3 | 377 | |||
D11327.1 | - | 4 | 481 | |||
M64322.1 | - | 4 | 469 |