Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271899.1 | NP_001258828.1 | 4 | 512 | ||
NM_001271900.1 | NP_001258829.1 | 5 | 706 | |||
NM_001271948.1 | NP_001258877.1 | 4 | 1039 | |||
NM_181674.2 | NP_858060.2 | 4 | 837 | |||
NM_181675.3 | NP_858061.2 | 4 | 559 | |||
NM_181676.2 | NP_858062.1 | 3 | 643 | |||
NM_181677.2 | NP_858063.1 | 5 | 469 | |||
NM_181678.2 | NP_858064.1 | 5 | 482 | |||
NR_073526.1 | 3 | 954 | ||||
NR_073527.1 | 4 | 591 | ||||
GenBank mRNA | AK056192.1 | - | 4 | 519 | ||
AK289717.1 | - | 4 | 488 | |||
AK294659.1 | - | 5 | 615 | |||
AK295347.1 | - | 4 | 483 | |||
AK314810.1 | - | 4 | 836 | |||
AL540315.3 | - | 2 | 172 | |||
AV726282.1 | - | 5 | 388 | |||
BC031790.1 | - | 4 | 727 | 4939981 | ||
BI669304.1 | - | 5 | 467 | |||
BX647887.1 | - | 4 | 572 | |||
DA155307.1 | - | 5 | 448 | |||
DA365141.1 | - | 5 | 469 | |||
M64930.1 | - | 3 | 765 |