Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000228.2 | NP_000219.2 | 10 | 1206 | ||
NM_001017402.1 | NP_001017402.1 | 9 | 1452 | |||
NM_001127641.1 | NP_001121113.1 | 10 | 1180 | |||
XM_005273124.4 | XP_005273181.1 | 10 | 1388 | |||
XM_017001272.1 | XP_016856761.1 | 9 | 1616 | |||
GenBank mRNA | AK296851.1 | - | 10 | 1156 | ||
AY035783.1 | - | 9 | 1062 | |||
BC075838.1 | - | 9 | 1237 | 6291096 | ||
D37766.1 | - | 10 | 1143 | |||
KU178159.1 | - | 9 | 1062 | |||
L25541.1 | - | 10 | 1179 |