Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000230.2 | NP_000221.1 | 3 | 768 | ||
XM_005250340.4 | XP_005250397.1 | 3 | 842 | |||
GenBank mRNA | BC060830.1 | - | 3 | 766 | 30333193 | |
U43653.1 | - | 2 | 767 |