Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000239.2 | NP_000230.1 | 2 | 241 | ||
GenBank mRNA | AK130149.1 | - | 2 | 246 | ||
AK310930.1 | - | 223 | ||||
AK311779.1 | - | 2 | 214 | |||
BC004147.2 | - | 2 | 205 | 2959387 | ||
FJ795023.1 | - | 2 | 208 | |||
J03801.1 | - | 2 | 211 | |||
KJ841928.1 | - | 2 | 186 | |||
M19045.1 | - | 2 | 199 | |||
M21119.1 | - | 2 | 199 | |||
U25677.1 | - | 2 | 186 |