Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000239.2 | NP_000230.1 | 4 | 496 | ||
GenBank mRNA | AK130149.1 | - | 4 | 501 | ||
AK310930.1 | - | 378 | ||||
BC004147.2 | - | 4 | 460 | 2959387 | ||
FJ795023.1 | - | 4 | 463 | |||
J03801.1 | - | 4 | 466 | |||
M19045.1 | - | 4 | 454 | |||
M21119.1 | - | 4 | 454 |