Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000283.3 | NP_000274.2 | 2 | 646 | ||
NM_001145291.1 | NP_001138763.1 | 2 | 646 | |||
XM_011513473.2 | XP_011511775.1 | 3 | 1913 | |||
XM_011513474.2 | XP_011511776.1 | 3 | 1913 | |||
XM_011513475.2 | XP_011511777.1 | 5 | 2016 | |||
XM_011513476.2 | XP_011511778.1 | 3 | 1913 | |||
GenBank mRNA | BC000249.1 | - | 2 | 636 | 3356353 | |
BT009794.1 | - | 2 | 593 | |||
S41458.1 | - | 2 | 614 | |||
X66142.1 | - | 2 | 598 |