Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000283.3 | NP_000274.2 | 6, 7 | 1045 | ||
NM_001145291.1 | NP_001138763.1 | 6, 7 | 1045 | |||
NM_001145292.1 | NP_001138764.1 | 4, 5 | 424 | |||
XM_011513473.2 | XP_011511775.1 | 7, 8 | 2312 | |||
XM_011513474.2 | XP_011511776.1 | 7, 8 | 2312 | |||
XM_011513475.2 | XP_011511777.1 | 9, 10 | 2415 | |||
XM_011513476.2 | XP_011511778.1 | 7, 8 | 2312 | |||
XM_011513478.2 | XP_011511780.1 | 3, 4 | 1231 | |||
XM_017008283.1 | XP_016863772.1 | 4, 5 | 451 | |||
XM_017008284.1 | XP_016863773.1 | 4, 5 | 406 | |||
XM_017008285.1 | XP_016863774.1 | 4, 5 | 577 | |||
XM_017008286.1 | XP_016863775.1 | 4, 5 | 406 | |||
GenBank mRNA | AK295285.1 | - | 4, 5 | 421 | ||
AK316054.1 | - | 4, 5 | 381 | |||
BC000249.1 | - | 6, 7 | 1035 | 3356353 | ||
BT009794.1 | - | 6, 7 | 992 | |||
S41458.1 | - | 6, 7 | 1013 | |||
X66142.1 | - | 6, 7 | 997 |