Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000285.3 | NP_000276.2 | 9, 10 | 812 | ||
NM_001166057.1 | NP_001159529.1 | 7, 8 | 620 | |||
GenBank mRNA | AK225037.1 | - | 9, 10 | 693 | ||
AK290756.1 | - | 9, 10 | 693 | |||
AK290781.1 | - | 9, 10 | 708 | |||
AK291561.1 | - | 9, 10 | 710 | |||
AK293126.1 | - | 7, 8 | 515 | |||
BC004305.1 | - | 9, 10 | 683 | 3626134 | ||
BC015027.1 | - | 9, 10 | 713 | 3920275 | ||
BC028295.1 | - | 9, 10 | 684 | 4470066 | ||
BT006692.1 | - | 9, 10 | 679 | |||
CR541669.1 | - | 9, 10 | 679 | |||
J04605.1 | - | 9, 10 | 695 |