Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000286.2 | NP_000277.1 | 2 | 1084 | ||
GenBank mRNA | AB004546.1 | - | 2 | 653 | ||
AK312635.1 | - | 2 | 997 | |||
BC015751.1 | - | 2 | 999 | |||
BC031085.1 | - | 2 | 988 | 5296541 | ||
U91521.1 | - | 2 | 1015 |