Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000287.3 | NP_000278.3 | 17 | 3208 | ||
NM_001316313.1 | NP_001303242.1 | 17 | 2944 | |||
NR_133009.1 | 15 | 2961 | ||||
XM_011514661.1 | XP_011512963.1 | 16 | 3124 | |||
GenBank mRNA | AB051076.1 | - | 17 | 3115 | ||
AB208906.1 | - | 13 | 3802 | |||
BC033487.1 | - | 17 | 3123 | |||
BC048331.1 | - | 17 | 3131 | 5189685 | ||
D83703.1 | - | 17 | 3170 |