Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000291.3 | NP_000282.1 | 11 | 1936 | ||
GenBank mRNA | AB062432.1 | - | 11 | 1833 | ||
BC104837.1 | - | 11 | 1814 | 8143840 | ||
BC113568.1 | - | 11 | 1815 | 8322620 |