Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000293.2 | NP_000284.1 | 7, 8 | 759 | ||
NM_001031835.2 | NP_001027005.1 | 8, 9 | 871 | |||
XM_005255983.4 | XP_005256040.1 | 7, 8 | 722 | |||
XM_005255984.4 | XP_005256041.1 | 8, 9 | 1022 | |||
XM_017023283.1 | XP_016878772.1 | 8, 9 | 844 | |||
XM_017023284.1 | XP_016878773.1 | 7, 8 | 725 | |||
GenBank mRNA | AK309226.1 | - | 7, 8 | 722 | ||
BC033657.1 | - | 8, 9 | 850 | 3913693 | ||
CR936710.1 | - | 8, 9 | 905 | |||
X84908.1 | - | 7, 8 | 731 |