Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000293.2 | NP_000284.1 | 31 | 3421 | ||
NM_001031835.2 | NP_001027005.1 | 32 | 3533 | |||
XM_005255983.4 | XP_005256040.1 | 31 | 3384 | |||
XM_005255984.4 | XP_005256041.1 | 32 | 3684 | |||
XM_017023282.1 | XP_016878771.1 | 25 | 2678 | |||
XM_017023283.1 | XP_016878772.1 | 33 | 3589 | |||
XM_017023284.1 | XP_016878773.1 | 32 | 3470 | |||
GenBank mRNA | AK123692.1 | - | 7 | 1705 | ||
AK293122.1 | - | 13 | 1541 | |||
BC033657.1 | - | 32 | 3512 | 3913693 | ||
CR936710.1 | - | 32 | 3565 | |||
X84908.1 | - | 31 | 3393 |