Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000304.3 | NP_000295.1 | 5 | 692 | ||
NM_001281455.1 | NP_001268384.1 | 5 | 653 | |||
NM_001281456.1 | NP_001268385.1 | 5 | 688 | |||
NM_153321.2 | NP_696996.1 | 5 | 649 | |||
NM_153322.2 | NP_696997.1 | 4 | 571 | |||
NR_104017.1 | 4 | 580 | ||||
NR_104018.1 | 3 | 480 | ||||
GenBank mRNA | AK290640.1 | - | 5 | 602 | ||
AK300690.1 | - | 6 | 511 | |||
BC019040.2 | - | 5 | 614 | 4588473 | ||
BC091499.1 | - | 5 | 609 | 30395315 | ||
BQ212717.1 | - | 1 | 52 | |||
BQ232542.1 | - | 3 | 361 | |||
CD519144.1 | - | 1 | 146 | |||
CR541931.1 | - | 4 | 454 | |||
CR541953.1 | - | 4 | 454 | |||
D11428.1 | - | 5 | 642 | |||
KR259962.1 | - | 3 | 436 | |||
L03203.1 | - | 5 | 568 | |||
M94048.1 | - | 4 | 503 | |||
X65968.1 | - | 2 | 275 |