Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004223.4 | NP_004214.1 | 4 | 721 | ||
NM_198183.2 | NP_937826.1 | 4 | 1080 | |||
GenBank mRNA | AF061736.1 | - | 4 | 672 | ||
AK093462.1 | - | 4 | 1034 | |||
AK129621.1 | - | 4 | 592 | |||
AK226165.1 | - | 4 | 775 | |||
BC032491.1 | - | 4 | 721 | 5246109 |