Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000905.3 | NP_000896.1 | 4 | 453 | ||
GenBank mRNA | BC029497.1 | - | 4 | 453 | 5278692 | |
K01911.1 | - | 4 | 451 | |||
M15789.1 | - | 3 | 378 |