Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002268.4 | NP_002259.1 | 17 | 2319 | ||
GenBank mRNA | AK129841.1 | - | 1 | 19 | ||
AK291041.1 | - | 17 | 2271 | |||
BC007458.1 | - | 1 | 209 | |||
BC016754.1 | - | 1 | 232 | |||
BC028691.2 | - | 17 | 2264 | 4838709 | ||
BC034493.2 | - | 17 | 2319 | 4821602 |