Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002345.3 | NP_002336.1 | 3 | 1607 | ||
GenBank mRNA | AK223177.1 | - | 3 | 1292 | ||
BC007038.1 | - | 3 | 1359 | 3950745 | ||
BC035997.1 | - | 3 | 1281 | 4688606 | ||
U18728.1 | - | 3 | 1298 | |||
U21128.1 | - | 3 | 1302 |