Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111097.2 | NP_001104567.1 | 7 | 841 | ||
NM_002350.3 | NP_002341.1 | 7 | 904 | |||
XM_011517529.2 | XP_011515831.1 | 6 | 3289 | |||
XM_017013415.1 | XP_016868904.1 | 6 | 3289 | |||
XM_017013416.1 | XP_016868905.1 | 7 | 580 | |||
GenBank mRNA | AK290494.1 | - | 7 | 814 | ||
AK298677.1 | - | 5 | 492 | |||
BC059394.1 | - | 7 | 833 | |||
BC068551.1 | - | 7 | 833 | |||
BC075001.2 | - | 6 | 647 | 30915399 | ||
BC075002.2 | - | 6 | 647 | 30915611 | ||
BC126456.1 | - | 8 | 741 | 8992172 | ||
BC126458.1 | - | 8 | 788 | 8992174 | ||
M16038.1 | - | 7 | 919 | |||
M79321.1 | - | 6 | 583 |