Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004252.4 | NP_004243.1 | 3 | 941 | ||
GenBank mRNA | AF015926.1 | - | 3 | 918 | ||
AF036241.1 | - | 3 | 876 | |||
AK026581.1 | - | 3 | 924 | |||
AK128474.1 | - | 4 | 890 | |||
AK312498.1 | - | 3 | 957 | |||
BC001443.2 | - | 3 | 894 | 3138881 | ||
BC003361.1 | - | 3 | 911 | 2900516 | ||
BC011777.1 | - | 3 | 910 | 3640162 | ||
BC049220.1 | - | 3 | 917 | |||
BC053350.1 | - | 3 | 901 | 5218414 |