Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002354.2 | NP_002345.2 | 5 | 870 | ||
GenBank mRNA | AK026585.1 | - | 5 | 707 | ||
BC014785.1 | - | 5 | 694 | 3861826 | ||
CR542259.1 | - | 5 | 512 | |||
CR542283.1 | - | 5 | 512 | |||
M26481.1 | - | 5 | 671 | |||
M32306.1 | - | 5 | 533 | |||
M32325.1 | - | 5 | 666 | |||
M33011.1 | - | 5 | 619 | |||
X14758.1 | - | 5 | 666 |