Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002356.6 | NP_002347.5 | 2 | 2584 | ||
GenBank mRNA | AK074526.1 | - | 2 | 1568 | ||
BC089040.1 | - | 2 | 2432 | 6378349 | ||
D10522.1 | - | 2 | 2541 |