Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184732.1 | NP_001171661.1 | 2 | 344 | ||
NM_004254.3 | NP_004245.2 | 2 | 365 | |||
GenBank mRNA | AB042505.1 | - | 2 | 347 | ||
AF097491.1 | - | 1 | 223 | |||
AK298351.1 | - | 3 | 317 | |||
AK313187.1 | - | 2 | 338 | |||
BC022387.1 | - | 2 | 359 | 4608542 | ||
DA628705.1 | - | 2 | 365 | |||
DA633836.1 | - | 2 | 314 |