Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002364.4 | NP_002355.2 | 2 | 431 | ||
XM_011545512.1 | XP_011543814.1 | 2 | 471 | |||
GenBank mRNA | AF015766.1 | - | 2 | 429 | ||
AK223081.1 | - | 2 | 413 | |||
AK313347.1 | - | 2 | 427 | |||
BC026071.2 | - | 2 | 426 | 4831169 |